Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(difluoromethoxy)naphthalene ≥97% research grade

Supplier: AOB CHEM USA
Warning

36388-1G 36388-25G 36388-500MG 36388-5G
76888-392EA 244.01 USD InStock
76888-392 76888-394 76888-396 76888-398
1-Bromo-4-(difluoromethoxy)naphthalene ≥97% research grade
1-Bromo-4-(difluoromethoxy)naphthalene


Formula: C₁₁H₇BrF₂O
MW: 273.073 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18410319
CAS Number: 1261783-62-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty