Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,2,4]Triazolo[4,3-a]pyridin-8-ylmethanol ≥95%

Supplier: AOB CHEM USA
Warning

36950-1G 36950-500MG 36950-5G
76888-764EA 510.3 USD InStock
76888-764 76888-766 76888-768
[1,2,4]Triazolo[4,3-a]pyridin-8-ylmethanol ≥95%
[1,2,4]Triazolo[4,3-a]pyridin-8-ylmethanol


Formula: C₇H₇N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD26939079
CAS Number: 1823349-14-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty