Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloro-6-(trifluoromethyl)phenyl)(morpholino)methanone ≥97% research grade

Supplier: AOB CHEM USA
Warning

37204-1G 37204-25G 37204-500MG 37204-5G
76888-898EA 140.04 USD InStock
76888-898 76888-900 76888-902 76888-904
(2-Chloro-6-(trifluoromethyl)phenyl)(morpholino)methanone ≥97% research grade
(2-Chloro-6-(trifluoromethyl)phenyl)(morpholino)methanone


Formula: C₁₂H₁₁ClF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632161
CAS Number: 2379322-46-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty