Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethoxy-2,3-difluoro-4-iodobenzene ≥97%

Supplier: AOB CHEM USA
Warning

37458-1G 37458-25G 37458-500MG 37458-5G
76888-996EA 263.11 USD InStock
76888-996 76888-998 76889-000 76889-002
1-Ethoxy-2,3-difluoro-4-iodobenzene ≥97%
1-Ethoxy-2,3-difluoro-4-iodobenzene


Formula: C₈H₇F₂IO
Storage Temperature: Ambient
MDL Number: MFCD16619252
CAS Number: 144292-42-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty