Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Trichloro-2-ethoxybenzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

37957-1G 37957-25G 37957-500MG 37957-5G
76889-242EA 233.4 USD InStock
76889-242 76889-244 76889-246 76889-248
1,3,5-Trichloro-2-ethoxybenzene ≥97% research grade
1,3,5-Trichloro-2-ethoxybenzene


Formula: C₈H₇Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD29044109
CAS Number: 23399-88-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty