Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-ethyl 3-(4-chloro-2-(trifluoromethyl)phenyl)acrylate ≥97%

Supplier: AOB CHEM USA
Warning

38138-1G 38138-25G 38138-5G 38138-500MG
76889-370EA 96.55 USD InStock
76889-370 76889-372 76889-374 77646-544
(E)-ethyl 3-(4-chloro-2-(trifluoromethyl)phenyl)acrylate ≥97%
(E)-ethyl 3-(4-chloro-2-(trifluoromethyl)phenyl)acrylate


Formula: C₁₂H₁₀ClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD23379725
CAS Number: 2413446-67-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty