Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trifluoro-1-(3-methoxyphenyl)ethanol ≥97%

Supplier: AOB CHEM USA
Warning

38795-1G 38795-25G 38795-500MG 38795-5G
76889-722EA 272.66 USD InStock
76889-722 76889-724 76889-726 76889-728
2,2,2-Trifluoro-1-(3-methoxyphenyl)ethanol ≥97%
2,2,2-Trifluoro-1-(3-methoxyphenyl)ethanol


Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00233113
CAS Number: 128816-77-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty