Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[4-Methanesulfonyl-3-(Trifluoromethyl)Phenyl]Boronic Acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

42773-1G 42773-25G 42773-500MG 42773-5G
76889-882EA 346.92 USD InStock
76889-882 76889-884 76889-886 76889-888
[4-Methanesulfonyl-3-(Trifluoromethyl)Phenyl]Boronic Acid ≥95% research grade
[4-Methanesulfonyl-3-(Trifluoromethyl)Phenyl]Boronic Acid


Formula: C₈H₈BF₃O₄S
Storage Temperature: Ambient
MDL Number: MFCD27935820
CAS Number: 1417301-54-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty