Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-((4-Bromophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOB CHEM USA
Warning

45625-1G 45625-25G 45625-500MG 45625-5G
76890-000EA 352.22 USD InStock
76890-000 76890-002 76890-004 76890-006
2-(4-((4-Bromophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-((4-Bromophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane


Formula: C₁₉H₂₂BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD31916459
CAS Number: 2246545-69-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty