Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-(Benzyloxy)-2-fluorophenyl)methanol ≥95% research grade

Supplier: AOB CHEM USA
Warning

46150-1G 46150-25G 46150-500MG 46150-5G
76890-122EA 162.32 USD InStock
76890-122 76890-124 76890-126 76890-128
(5-(Benzyloxy)-2-fluorophenyl)methanol ≥95% research grade
(5-(Benzyloxy)-2-fluorophenyl)methanol


Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707161
CAS Number: 459215-46-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty