Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Bromo-2-fluoro-6-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOB CHEM USA
Warning

48743-1G 48743-25G 48743-500MG 48743-5G
76890-590EA 314.03 USD InStock
76890-590 76890-592 76890-594 76890-596
2-(3-Bromo-2-fluoro-6-methylphenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-2-fluoro-6-methylphenyl)-1,3-dioxolane


Formula: C₁₀H₁₀BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD33403902
CAS Number: 2635937-52-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty