Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Difluoro-4-iodo-3-methoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

48821-1G 48821-25G 48821-500MG 48821-5G
76890-638EA 177.18 USD InStock
76890-638 76890-640 76890-642 76890-644
1,2-Difluoro-4-iodo-3-methoxybenzene ≥97%
1,2-Difluoro-4-iodo-3-methoxybenzene


Formula: C₇H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD32902774
CAS Number: 2091145-96-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty