Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-5-fluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

49422-1G 49422-25G 49422-500MG 49422-5G
76890-736EA 340.55 USD InStock
76890-736 76890-738 76890-740 76890-742
2,3-Dichloro-5-fluorobenzoic acid ≥97%
2,3-Dichloro-5-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845992
CAS Number: 1160573-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty