Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromophenyl)-2,2-difluoroethanone ≥95% research grade

Supplier: AOB CHEM USA
Warning

52570-1G 52570-25G 52570-500MG 52570-5G
76891-276EA 340.55 USD InStock
76891-276 76891-278 76891-280 76891-282
1-(4-Bromophenyl)-2,2-difluoroethanone ≥95% research grade
1-(4-Bromophenyl)-2,2-difluoroethanone


Formula: C₈H₅BrF₂O
MW: 235.026 g/mol
Storage Temperature: Refrigerator
CAS Number: 173974-88-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty