Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-difluorobenzyl alcohol ≥97% research grade

Supplier: AOB CHEM USA
Warning

53434-1G 53434-25G 53434-500MG 53434-5G
76891-446EA 439.22 USD InStock
76891-446 76891-448 76891-450 76891-452
4-Bromo-3,5-difluorobenzyl alcohol ≥97% research grade
4-Bromo-3,5-difluorobenzyl alcohol


Formula: C₇H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22381218
CAS Number: 1256276-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty