Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Bromobenzo[b]thiophen-2-yl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

54342-1G 54342-25G 54342-500MG 54342-5G
76891-582EA 173.99 USD InStock
76891-582 76891-584 76891-586 76891-588
(3-Bromobenzo[b]thiophen-2-yl)boronic acid ≥97% research grade
(3-Bromobenzo[b]thiophen-2-yl)boronic acid


Formula: C₈H₆BBrO₂S
MW: 256.912 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34530254
CAS Number: 2488711-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty