Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-6-methoxypyridine ≥97%

Supplier: AOB CHEM USA
Warning

55390-25G 55390-5G
76891-784EA 616.39 USD InStock
76891-784 76891-786
2,3-Dibromo-6-methoxypyridine ≥97%
2,3-Dibromo-6-methoxypyridine


Formula: C₆H₅Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD21648443
CAS Number: 909720-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty