Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Hydroxycyclohexyl)acetic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

55572-1G 55572-25G 55572-500MG 55572-5G
76891-810EA 155.96 USD InStock
76891-810 76891-812 76891-814 76891-816
(1-Hydroxycyclohexyl)acetic acid ≥95% research grade
(1-Hydroxycyclohexyl)acetic acid


Formula: C₈H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01028654
CAS Number: 14399-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty