Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-5-phenyl-1H-pyrazole ≥97% research grade

Supplier: AOB CHEM USA
Warning

56169-1G 56169-25G 56169-500MG 56169-5G
76891-868EA 188.85 USD InStock
76891-868 76891-870 76891-872 76891-874
1-Methyl-5-phenyl-1H-pyrazole ≥97% research grade
1-Methyl-5-phenyl-1H-pyrazole


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14643706
CAS Number: 3463-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty