Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isobutyl-1H-pyrazole-5-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

56548-1G 56548-25G 56548-500MG 56548-5G
76891-894EA 170.81 USD InStock
76891-894 76891-896 76891-898 76891-900
1-Isobutyl-1H-pyrazole-5-carboxylic acid ≥97%
1-Isobutyl-1H-pyrazole-5-carboxylic acid


Formula: C₈H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD6805234
CAS Number: 1006493-85-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty