Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Bromo-2-chloro-4-fluorophenyl)methanol ≥97%

Supplier: AOB CHEM USA
Warning

58601-1G 58601-25G 58601-500MG 58601-5G
76892-170EA 367.08 USD InStock
76892-170 76892-172 76892-174 76892-176
(5-Bromo-2-chloro-4-fluorophenyl)methanol ≥97%
(5-Bromo-2-chloro-4-fluorophenyl)methanol


Formula: C₇H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD31621013
CAS Number: 1313512-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty