Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Aminothiazol-5-yl)-4-bromophenol ≥95% research grade

Supplier: AOB CHEM USA
Warning

62858-1G 62858-25G 62858-500MG 62858-5G
76892-644EA 436.03 USD InStock
76892-644 76892-646 76892-648 76892-650
2-(2-Aminothiazol-5-yl)-4-bromophenol ≥95% research grade
2-(2-Aminothiazol-5-yl)-4-bromophenol


Formula: C₉H₇BrN₂OS
MW: 271.1336 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-75-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty