Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthyl trifluoromethyl ketone ≥97%

Supplier: AOB CHEM USA
Warning

64112-1G 64112-25G 64112-500MG 64112-5G
76892-866EA 144.29 USD InStock
76892-866 76892-868 76892-870 76892-872
2-Naphthyl trifluoromethyl ketone ≥97%
2-Naphthyl trifluoromethyl ketone


Formula: C₁₂H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD01320008
CAS Number: 1800-42-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty