Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-fluoropyridine ≥97%

Supplier: AOB CHEM USA
Warning

68925-1G 68925-25G 68925-500MG 68925-5G
76893-198EA 161.26 USD InStock
76893-198 76893-200 76893-202 76893-204
2,4-Dichloro-5-fluoropyridine ≥97%
2,4-Dichloro-5-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂Cl₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD18257011
CAS Number: 189281-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty