Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4-formylbenzonitrile 95%

Supplier: AOB CHEM USA
Warning

69769-1G 69769-500MG 69769-5G 69769-25G
76893-250EA 1544.68 USD InStock
76893-250 76893-252 77633-690 77656-824
2,3-Difluoro-4-formylbenzonitrile 95%
2,3-Difluoro-4-formylbenzonitrile


Formula: C₈H₃F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD24684755
CAS Number: 717883-41-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty