Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol ≥97% research grade

Supplier: AOB CHEM USA
Warning

75715-1G 75715-500MG 75715-5G 75715-25G
76893-458EA 690.65 USD InStock
76893-458 76893-460 76893-462 77660-324
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol ≥97% research grade
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutanol


Formula: C₁₆H₂₃BO₃
MW: 274.163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18734557
CAS Number: 1398331-98-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty