Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Amino-3-methylphenyl)ethanol ≥97%

Supplier: AOB CHEM USA
Warning

79425-1G 79425-25G 79425-500MG 79425-5G
76893-608EA 337.37 USD InStock
76893-608 76893-610 76893-612 76893-614
1-(2-Amino-3-methylphenyl)ethanol ≥97%
1-(2-Amino-3-methylphenyl)ethanol


Formula: C₉H₁₃NO
Storage Temperature: Refrigerator
CAS Number: 196611-19-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty