Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-Chloro-4-(Methylsulfonyl)Benzene ≥97% research grade

Supplier: AOB CHEM USA
Warning

91164-1G 91164-25G 91164-500MG 91164-5G
76893-888EA 377.69 USD InStock
76893-888 76893-890 76893-892 76893-894
1-Bromo-2-Chloro-4-(Methylsulfonyl)Benzene ≥97% research grade
1-Bromo-2-Chloro-4-(Methylsulfonyl)Benzene


Formula: C₇H₆BrClO₂S
MW: 269.543 g/mol
Storage Temperature: Ambient
CAS Number: 1346555-48-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty