Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Bromophenoxy)-2-methyl-4-nitrobenzene ≥97%

Supplier: AOB CHEM USA
Warning

94019-1G 94019-25G 94019-500MG 94019-5G
76893-956EA 145.35 USD InStock
76893-956 76893-958 76893-960 76893-962
1-(2-Bromophenoxy)-2-methyl-4-nitrobenzene ≥97%
1-(2-Bromophenoxy)-2-methyl-4-nitrobenzene


Formula: C₁₃H₁₀BrNO₃
Storage Temperature: Ambient
CAS Number: 1152822-02-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty