Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,3,5-trimethoxybenzene ≥97%

Supplier: AOB CHEM USA
Warning

99335-1G 99335-500MG 99335-5G
76893-996EA 738.39 USD InStock
76893-996 76893-998 76894-000
1-Bromo-2,3,5-trimethoxybenzene ≥97%
1-Bromo-2,3,5-trimethoxybenzene


Formula: C₉H₁₁BrO₃
Storage Temperature: Refrigerator
CAS Number: 23030-39-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty