Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

10150-1G 10150-25G 10150-500MG 10150-5G
76906-278EA 124.13 USD InStock
76906-278 76906-280 76906-282 76906-284
4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid ≥97% research grade
4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid


Formula: C₁₆H₂₀BN₃O₃
MW: 313.159 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08689618
CAS Number: 1309980-72-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty