Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

10493-25G 10493-5G
76906-416EA 310.85 USD InStock
76906-416 76906-418
3-Fluoro-4-pyridineboronic acid pinacol ester ≥97%
3-Fluoro-4-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₅BFNO₂
MW: 223.05
Storage Temperature: Refrigerator
MDL Number: MFCD06798251
CAS Number: 458532-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty