Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dinitropyridin-4-ol ≥97%

Supplier: AOB CHEM USA
Warning

107966-1G 107966-25G 107966-500MG 107966-5G
76906-502EA 212.18 USD InStock
76906-502 76906-504 76906-506 76906-508
3,5-Dinitropyridin-4-ol ≥97%
3,5-Dinitropyridin-4-ol


Formula: C₅H₃N₃O₅
Storage Temperature: Refrigerator
CAS Number: 10425-71-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty