Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formyl-4-thiopheneboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

12047-1G 12047-25G 12047-5G
76906-878EA 84.88 USD InStock
76906-878 76906-880 76906-882
3-Formyl-4-thiopheneboronic acid ≥97%
3-Formyl-4-thiopheneboronic acid
Precaución: for Research Use Only


Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01319006
CAS Number: 4347-32-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty