Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(furan-3-yl)benzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

122077-1G 122077-500MG 122077-5G 122077-25G
76906-938EA 622.75 USD InStock
76906-938 76906-940 76906-942 77653-362
3-(furan-3-yl)benzaldehyde ≥97% research grade
3-(furan-3-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD19612477
CAS Number: 263349-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty