Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluoro-4-pyridineboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

13562-1G 13562-25G 13562-5G
76907-210EA 84.88 USD InStock
76907-210 76907-212 76907-214
3-Chloro-2-fluoro-4-pyridineboronic acid ≥97%
3-Chloro-2-fluoro-4-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₅H₄BClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09037477
CAS Number: 1217500-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty