Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carboxy-2-methoxyphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

13608-1G 13608-25G 13608-500MG 13608-5G
76907-246EA 142.17 USD InStock
76907-246 76907-248 76907-250 76907-252
3-Carboxy-2-methoxyphenylboronic acid ≥97%
3-Carboxy-2-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₉BO₅
Storage Temperature: Refrigerator
MDL Number: MFCD09027237
CAS Number: 913836-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty