Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carbamoyl-3-chlorobenzeneboronic acid ≥95%

Supplier: AOB CHEM USA
Warning

15372-1G 15372-25G 15372-500MG 15372-5G
76907-918EA 141.1 USD InStock
76907-918 76907-920 76907-922 76907-924
4-Carbamoyl-3-chlorobenzeneboronic acid ≥95%
4-Carbamoyl-3-chlorobenzeneboronic acid


Formula: C₇H₇BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07363777
CAS Number: 850589-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty