Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Formyl-3,5-dimethylphenyl)boronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

16693-1G 16693-25G 16693-500MG 16693-5G
76908-588EA 147.47 USD InStock
76908-588 76908-590 76908-592 76908-594
(4-Formyl-3,5-dimethylphenyl)boronic acid ≥97% research grade
(4-Formyl-3,5-dimethylphenyl)boronic acid


Formula: C₉H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD15143475
CAS Number: 1218790-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty