Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-methoxy-5-trifluoromethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

16827-1G 16827-25G 16827-500MG 16827-5G
76908-644EA 203.7 USD InStock
76908-644 76908-646 76908-648 76908-650
3-Fluoro-4-methoxy-5-trifluoromethylphenylboronic acid ≥97%
3-Fluoro-4-methoxy-5-trifluoromethylphenylboronic acid


Formula: C₈H₇BF₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD16295242
CAS Number: 1451391-99-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty