Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylsulfonyl)-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

17319-1G 17319-25G 17319-500MG 17319-5G
76908-876EA 153.84 USD InStock
76908-876 76908-878 76908-880 76908-882
4-(Methylsulfonyl)-2-(trifluoromethyl)phenylboronic acid ≥97%
4-(Methylsulphonyl)-2-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BF₃O₄S
Storage Temperature: Ambient
MDL Number: MFCD10699623
CAS Number: 1072946-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty