Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-ethoxy-5-methylbenzaldehyde ≥97% research grade

Supplier: AOB CHEM USA
Warning

18187-1G 18187-25G 18187-500MG 18187-5G
76909-208EA 171.87 USD InStock
76909-208 76909-210 76909-212 76909-214
3-Bromo-2-ethoxy-5-methylbenzaldehyde ≥97% research grade
3-Bromo-2-ethoxy-5-methylbenzaldehyde


Formula: C₁₀H₁₁BrO₂
MW: 243.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105357
CAS Number: 708272-19-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty