Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Carboxy-5-methylphenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

19009-1G 19009-25G 19009-500MG 19009-5G
76909-540EA 350.1 USD InStock
76909-540 76909-542 76909-544 76909-546
3-Carboxy-5-methylphenylboronic acid ≥97% research grade
3-Carboxy-5-methylphenylboronic acid


Formula: C₈H₉BO₄
MW: 179.966 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025977
CAS Number: 1150114-66-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty