Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,3-dimethylphenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

19502-1G 19502-25G 19502-500MG 19502-5G
76909-738EA 225.98 USD InStock
76909-738 76909-740 76909-742 76909-744
4-Bromo-2,3-dimethylphenylboronic acid ≥97%
4-Bromo-2,3-dimethylphenylboronic acid


Formula: C₈H₁₀BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD16295270
CAS Number: 1451391-29-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty