Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Chlorobenzoyl)morpholine ≥97%

Supplier: AOB CHEM USA
Warning

19659-25G 19659-5G
76909-788EA 209 USD InStock
76909-788 76909-790
N-(4-Chlorobenzoyl)morpholine ≥97%
N-(4-Chlorobenzoyl)morpholine
Precaución: for Research Use Only


Formula: C₁₁H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00458710
CAS Number: 19202-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty