Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,6-dichlorophenylboronic acid ≥97% research grade

Supplier: AOB CHEM USA
Warning

20159-1G 20159-25G 20159-500MG 20159-5G
76909-960EA 173.99 USD InStock
76909-960 76909-962 76909-964 76909-966
3-Bromo-2,6-dichlorophenylboronic acid ≥97% research grade
3-Bromo-2,6-dichlorophenylboronic acid


Formula: C₆H₄BBrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD17011992
CAS Number: 1451392-94-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty