Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-isopropoxypyridine ≥97% research grade

Supplier: AOB CHEM USA
Warning

20492-1G 20492-25G 20492-500MG 20492-5G
76910-212EA 195.21 USD InStock
76910-212 76910-214 76910-216 76910-218
3-Chloro-2-isopropoxypyridine ≥97% research grade
3-Chloro-2-isopropoxypyridine


Formula: C₈H₁₀ClNO
MW: 171.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19689930
CAS Number: 282723-22-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty