Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)-5-(trifluoromethyl)phenylboronic acid ≥95% research grade

Supplier: AOB CHEM USA
Warning

21982-1G 21982-25G 21982-500MG 21982-5G
76910-680EA 190.97 USD InStock
76910-680 76910-682 76910-684 76910-686
3-(Hydroxymethyl)-5-(trifluoromethyl)phenylboronic acid ≥95% research grade
3-(Hydroxymethyl)-5-(trifluoromethyl)phenylboronic acid


Formula: C₈H₈BF₃O₃
MW: 219.954 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22398665
CAS Number: 2088638-48-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty