Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloropyridine-4-carboxylic acid ≥95%

Supplier: AOB CHEM USA
Warning

22533-1G 22533-25G 22533-5G
76910-858EA 84.88 USD InStock
76910-858 76910-860 76910-862
3-Bromo-2-chloropyridine-4-carboxylic acid ≥95%
3-Bromo-2-chloropyridine-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₃BrClNO₂
MW: 236.449 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185793
CAS Number: 1214323-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty