Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formyl-5-(trifluoromethyl)benzonitrile ≥97% research grade

Supplier: AOB CHEM USA
Warning

23501-1G 23501-25G 23501-500MG 23501-5G
76911-280EA 162.32 USD InStock
76911-280 76911-282 76911-284 76911-286
3-Formyl-5-(trifluoromethyl)benzonitrile ≥97% research grade
3-Formyl-5-(trifluoromethyl)benzonitrile


Formula: C₉H₄F₃NO
MW: 199.1294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544347
CAS Number: 569370-39-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty